The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005257641.4 | XP_005257698.1 | 1 - 2 | 889 | 118 |
GenBank mRNA | D13969.1 | - | 1 - 2 | 166 | 118 |