The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282902.2 | NP_001269831.1 | 4 - 5 | 2799 | 80 |
NM_001282903.2 | NP_001269832.1 | 5 - 6 | 3288 | 80 | |
NM_014112.4 | NP_054831.2 | 5 - 6 | 3302 | 80 | |
XM_005251049.2 | XP_005251106.1 | 4 - 5 | 3117 | 80 | |
XM_006716625.1 | XP_006716688.1 | 5 - 6 | 3239 | 80 | |
XM_011517264.1 | XP_011515566.1 | 6 - 7 | 3372 | 80 | |
XM_011517266.2 | XP_011515568.1 | 5 - 6 | 3107 | 80 | |
XM_011517267.1 | XP_011515569.1 | 5 - 6 | 3225 | 80 | |
XM_011517268.1 | XP_011515570.1 | 5 - 6 | 3214 | 80 | |
XM_017013801.1 | XP_016869290.1 | 4 - 5 | 6901 | 80 | |
GenBank mRNA | AF183810.1 | - | 5 - 6 | 3296 | 80 |
AF264784.1 | - | 4 - 5 | 2843 | 80 | |
AK000948.1 | - | 2 - 3 | 1286 | 80 | |
AK304046.1 | - | 4 - 5 | 2794 | 80 | |
BC125020.1 | - | 5 - 6 | 3132 | 80 |