The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014571.3 | NP_055386.1 | 1 - 2 | 131 | 78 |
GenBank mRNA | AF311885.1 | - | 1 - 2 | 80 | 78 |
AJ272215.1 | - | 1 - 2 | 90 | 78 | |
AL541019.3 | - | 1 - 2 | 131 | 78 | |
BC006087.1 | - | 1 - 2 | 101 | 78 |