The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001769.3 | NP_001760.1 | 1 - 2 | 248 | 72 |
XM_005253814.3 | XP_005253871.1 | 1 - 2 | 245 | 72 | |
GenBank mRNA | AB079244.1 | - | 1 - 2 | 169 | 72 |
AK296332.1 | - | 1 - 2 | 191 | 72 | |
AK296894.1 | - | 1 - 2 | 164 | 72 | |
AK308700.1 | - | 1 - 2 | 195 | 72 | |
AK315219.1 | - | 1 - 2 | 197 | 72 | |
AY423720.1 | - | 1 - 2 | 81 | 72 | |
AY966455.1 | - | 1 - 2 | 175 | 72 | |
BC011988.1 | - | 1 - 2 | 115 | 72 | |
BG291377.1 | - | 1 - 2 | 250 | 72 | |
BM763066.1 | - | 1 - 2 | 248 | 72 | |
L34068.1 | - | 1 - 2 | 64 | 72 | |
M38690.1 | - | 1 - 2 | 115 | 72 | |
X60111.1 | - | 1 - 2 | 175 | 72 |