Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ITGAE
Entrez Gene ID
3682
Gene Name
integrin subunit alpha E
Gene Aliases
CD103, HUMINAE
Chromosome Location
Chr.17: 3714625 - 3803766 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002208.4 NP_002199.3 16 - 17 2122 124
XM_011523823.2 XP_011522125.1 16 - 17 2560 124
XM_011523825.2 XP_011522127.1 16 - 17 2558 124
XM_011523827.2 XP_011522129.1 16 - 17 2552 124
XM_011523828.2 XP_011522130.1 16 - 17 2556 124
XM_017024586.1 XP_016880075.1 17 - 18 3945 124
XM_017024587.1 XP_016880076.1 17 - 18 3945 124
GenBank mRNA BC113436.1 - 16 - 17 2112 124
BC117207.1 - 16 - 17 2112 124
L25851.2 - 16 - 17 2148 124

View Full Product Details