Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SERPINB2
Entrez Gene ID
5055
Gene Name
serpin family B member 2
Gene Aliases
HsT1201, PAI, PAI-2, PAI2, PLANH2
Chromosome Location
Chr.18: 63887705 - 63903890 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143818.1 NP_001137290.1 3 - 4 501 120
NM_002575.2 NP_002566.1 2 - 3 243 120
GenBank mRNA A21238.1 - 2 - 3 216 120
A32415.1 - 2 - 3 209 120
AK313159.1 - 2 - 3 249 120
BC012609.1 - 2 - 3 247 120
DA543366.1 - 3 - 4 501 120
DQ656059.2 - 2 - 3 430 120
J02685.1 - 2 - 3 223 120
J03603.1 - 2 - 3 216 120
M18082.1 - 1 - 2 199 120
Y00630.1 - 2 - 3 240 120

View Full Product Details