The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242644.1 | NP_001229573.1 | 2 - 3 | 416 | 90 |
NM_002865.2 | NP_002856.1 | 3 - 4 | 488 | 90 | |
GenBank mRNA | AF498930.1 | - | 3 - 4 | 190 | 90 |
AK297576.1 | - | 2 - 3 | 390 | 90 | |
AK312344.1 | - | 3 - 4 | 411 | 90 | |
BC008929.2 | - | 3 - 4 | 378 | 90 | |
BT019695.1 | - | 3 - 4 | 190 | 90 | |
DA648682.1 | - | 3 - 4 | 490 | 90 | |
M28213.1 | - | 3 - 4 | 230 | 90 | |
X12953.1 | - | 3 - 4 | 398 | 90 |