Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 109605890-109606399 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNAT2
Entrez Gene ID
2780
Gene Name
G protein subunit alpha transducin 2
Gene Aliases
ACHM4, GNATC
UniGene
Hs.36973
Gene Chromosome Location
Chr.1: 109603091-109619733 on Build GRCh38
Transcripts
NM_005272.3 XM_011541264.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4687423 c.586T>G p.F196V Chr.1: 109606312-109606312 on Build GRCh38
5881480 c.686C>G p.A229G Chr.1: 109606004-109606004 on Build GRCh38
893544 c.627G>T p.K209N Chr.1: 109606063-109606063 on Build GRCh38
3470939 c.540A>T p.K180N Chr.1: 109606358-109606358 on Build GRCh38
893543 c.646G>A p.E216K Chr.1: 109606044-109606044 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1799940 Chr.1: 109606350-109606350 on Build GRCh38
rs566052246 Chr.1: 109605962-109605962 on Build GRCh38
rs187755113 Chr.1: 109606163-109606163 on Build GRCh38
rs374210835 Chr.1: 109606371-109606371 on Build GRCh38
rs772274756 Chr.1: 109605956-109605956 on Build GRCh38
rs748179055 Chr.1: 109605953-109605953 on Build GRCh38
rs143323029 Chr.1: 109606177-109606177 on Build GRCh38
rs909225562 Chr.1: 109606190-109606190 on Build GRCh38
rs995152260 Chr.1: 109606067-109606067 on Build GRCh38
rs551776784 Chr.1: 109606081-109606081 on Build GRCh38
rs751633351 Chr.1: 109606347-109606347 on Build GRCh38
rs534677249 Chr.1: 109606045-109606045 on Build GRCh38
rs74113953 Chr.1: 109606246-109606246 on Build GRCh38
rs746117122 Chr.1: 109606353-109606353 on Build GRCh38
rs1012677513 Chr.1: 109606286-109606286 on Build GRCh38
rs750089687 Chr.1: 109606318-109606318 on Build GRCh38
rs369242630 Chr.1: 109606148-109606148 on Build GRCh38
rs267597907 Chr.1: 109606071-109606071 on Build GRCh38
rs758844656 Chr.1: 109606333-109606333 on Build GRCh38
rs72987059 Chr.1: 109606142-109606142 on Build GRCh38
rs369719633 Chr.1: 109606116-109606116 on Build GRCh38
rs755458273 Chr.1: 109606060-109606060 on Build GRCh38
rs199666763 Chr.1: 109606108-109606108 on Build GRCh38
rs201798524 Chr.1: 109606348-109606348 on Build GRCh38
rs192670813 Chr.1: 109606265-109606265 on Build GRCh38
rs774443831 Chr.1: 109605931-109605931 on Build GRCh38
rs777780826 Chr.1: 109606262-109606262 on Build GRCh38
rs977295051 Chr.1: 109605915-109605915 on Build GRCh38
rs745663961 Chr.1: 109606278-109606278 on Build GRCh38
rs774099409 Chr.1: 109606363-109606364 on Build GRCh38
rs564109875 Chr.1: 109606189-109606189 on Build GRCh38
rs373111837 Chr.1: 109606271-109606271 on Build GRCh38
rs199969527 Chr.1: 109606272-109606272 on Build GRCh38
rs943921354 Chr.1: 109605918-109605918 on Build GRCh38
rs1048021529 Chr.1: 109606074-109606074 on Build GRCh38
rs998557563 Chr.1: 109606317-109606317 on Build GRCh38
rs113610946 Chr.1: 109606283-109606283 on Build GRCh38
rs932086888 Chr.1: 109606186-109606186 on Build GRCh38
rs1050400219 Chr.1: 109606001-109606001 on Build GRCh38
rs757273018 Chr.1: 109606146-109606147 on Build GRCh38
rs752857789 Chr.1: 109606176-109606176 on Build GRCh38
rs1799875 Chr.1: 109606352-109606352 on Build GRCh38
rs775349840 Chr.1: 109606123-109606123 on Build GRCh38
rs192819117 Chr.1: 109606133-109606133 on Build GRCh38
rs780429600 Chr.1: 109606042-109606042 on Build GRCh38
rs370869387 Chr.1: 109606344-109606344 on Build GRCh38
rs367863835 Chr.1: 109605976-109605976 on Build GRCh38
rs779949808 Chr.1: 109606372-109606372 on Build GRCh38
rs924446444 Chr.1: 109605917-109605917 on Build GRCh38
rs762529730 Chr.1: 109606125-109606125 on Build GRCh38
rs185290169 Chr.1: 109606145-109606145 on Build GRCh38
rs758050645 Chr.1: 109605989-109605989 on Build GRCh38
rs767182806 Chr.1: 109606302-109606302 on Build GRCh38
rs768739908 Chr.1: 109606290-109606290 on Build GRCh38
rs763825174 Chr.1: 109605992-109605992 on Build GRCh38
rs771116688 Chr.1: 109606269-109606269 on Build GRCh38
rs200705508 Chr.1: 109606102-109606102 on Build GRCh38
rs752187366 Chr.1: 109605982-109605982 on Build GRCh38
rs769768005 Chr.1: 109606118-109606118 on Build GRCh38
rs201230566 Chr.1: 109606018-109606018 on Build GRCh38
rs777323795 Chr.1: 109606211-109606211 on Build GRCh38
rs376412882 Chr.1: 109606328-109606328 on Build GRCh38
rs759910267 Chr.1: 109606327-109606327 on Build GRCh38
rs966699132 Chr.1: 109606242-109606242 on Build GRCh38
rs753083112 Chr.1: 109606332-109606332 on Build GRCh38
rs751307255 Chr.1: 109605995-109605995 on Build GRCh38
rs777936715 Chr.1: 109606076-109606076 on Build GRCh38
rs140231308 Chr.1: 109606072-109606072 on Build GRCh38
rs761511634 Chr.1: 109606141-109606141 on Build GRCh38
rs749110943 Chr.1: 109606373-109606373 on Build GRCh38
rs751150036 Chr.1: 109606136-109606136 on Build GRCh38
rs377096148 Chr.1: 109605961-109605961 on Build GRCh38
rs887755996 Chr.1: 109606007-109606007 on Build GRCh38
rs754223305 Chr.1: 109606143-109606143 on Build GRCh38
rs761188037 Chr.1: 109606117-109606117 on Build GRCh38
rs140788789 Chr.1: 109605975-109605975 on Build GRCh38
rs779352893 Chr.1: 109606063-109606063 on Build GRCh38
rs780043566 Chr.1: 109606198-109606198 on Build GRCh38
rs200962508 Chr.1: 109606291-109606291 on Build GRCh38
rs761237104 Chr.1: 109606299-109606299 on Build GRCh38
rs976644545 Chr.1: 109606172-109606172 on Build GRCh38
rs758934750 Chr.1: 109606257-109606257 on Build GRCh38
rs765099163 Chr.1: 109605979-109605979 on Build GRCh38
rs751935808 Chr.1: 109606188-109606188 on Build GRCh38
rs540640583 Chr.1: 109606187-109606187 on Build GRCh38
rs755321515 Chr.1: 109606207-109606207 on Build GRCh38
rs769526367 Chr.1: 109606280-109606280 on Build GRCh38
rs762948491 Chr.1: 109606288-109606288 on Build GRCh38
rs924592174 Chr.1: 109606160-109606160 on Build GRCh38

View Full Product Details