The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135599.2 | NP_001129071.1 | 7 - 8 | 2541 | 92 |
NM_003238.3 | NP_003229.1 | 6 - 7 | 2457 | 92 | |
GenBank mRNA | AK295671.1 | - | 7 - 8 | 905 | 92 |
AK296504.1 | - | 7 - 8 | 1174 | 92 | |
AK299839.1 | - | 6 - 7 | 651 | 92 | |
AK313106.1 | - | 6 - 7 | 1194 | 92 | |
BC096235.4 | - | 6 - 7 | 1243 | 92 | |
BC096236.4 | - | 6 - 7 | 1132 | 92 | |
BC099635.1 | - | 6 - 7 | 1247 | 92 | |
M19154.1 | - | 7 - 8 | 1640 | 92 | |
Y00083.1 | - | 6 - 7 | 1270 | 92 |