The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003635.3 | NP_003626.1 | 9 - 10 | 2647 | 70 |
XM_005270255.3 | XP_005270312.1 | 9 - 10 | 2647 | 70 | |
XM_005270256.3 | XP_005270313.1 | 6 - 7 | 852 | 70 | |
XM_011540310.2 | XP_011538612.1 | 8 - 9 | 3216 | 70 | |
XM_017016857.1 | XP_016872346.1 | 9 - 10 | 2676 | 70 | |
GenBank mRNA | AK293527.1 | - | 5 - 6 | 704 | 70 |
AK298957.1 | - | 7 - 8 | 940 | 70 | |
AK300520.1 | - | 5 - 6 | 850 | 70 | |
AK301251.1 | - | 4 - 5 | 529 | 70 | |
AK303648.1 | - | 10 - 11 | 2445 | 70 | |
AK310580.1 | - | 5 - 6 | 813 | 70 | |
BC035711.1 | - | 9 - 10 | 2400 | 70 | |
BC110588.1 | - | 7 - 8 | 1879 | 70 | |
BC110589.1 | - | 7 - 8 | 1879 | 70 | |
U36601.1 | - | 7 - 8 | 1870 | 70 |