The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004346.3 | NP_004337.2 | 4 - 5 | 441 | 100 |
NM_032991.2 | NP_116786.1 | 3 - 4 | 274 | 100 | |
XM_011532301.1 | XP_011530603.1 | 4 - 5 | 375 | 100 | |
GenBank mRNA | AJ413269.1 | - | 2 - 3 | 178 | 100 |
AK291337.1 | - | 4 - 5 | 412 | 100 | |
AU127028.1 | - | 3 - 4 | 274 | 100 | |
BC016926.2 | - | 4 - 5 | 393 | 100 | |
BX647609.1 | - | 4 - 5 | 513 | 100 | |
U13737.1 | - | 4 - 5 | 402 | 100 | |
U13738.1 | - | 3 - 4 | 100 | ||
U26943.1 | - | 2 - 3 | 178 | 100 |