The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001288729.1 | NP_001275658.1 | 18 - 19 | 2132 | 76 |
NM_001288730.1 | NP_001275659.1 | 18 - 19 | 2122 | 76 | |
NM_001288731.1 | NP_001275660.1 | 18 - 19 | 2105 | 76 | |
NM_004938.3 | NP_004929.2 | 18 - 19 | 2457 | 76 | |
XM_005251757.3 | XP_005251814.1 | 18 - 19 | 2227 | 76 | |
XM_017014360.1 | XP_016869849.1 | 5 - 6 | 1370 | 76 | |
GenBank mRNA | AB208871.1 | - | 18 - 19 | 2060 | 76 |
AK127855.1 | - | 4 - 5 | 2548 | 76 | |
AK295119.1 | - | 18 - 19 | 2189 | 76 | |
BC113660.1 | - | 17 - 18 | 1995 | 76 | |
BC143730.1 | - | 16 - 17 | 1945 | 76 | |
BC143733.1 | - | 17 - 18 | 1995 | 76 | |
BC143759.1 | - | 17 - 18 | 1995 | 76 | |
CR749834.1 | - | 18 - 19 | 2105 | 76 | |
DA855348.1 | - | 2 - 3 | 232 | 76 | |
X76104.1 | - | 18 - 19 | 2261 | 76 |