The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005894.2 | NP_005885.1 | 1 - 2 | 171 | 73 |
XM_005245602.1 | XP_005245659.1 | 1 - 2 | 171 | 73 | |
XM_017002806.1 | XP_016858295.1 | 1 - 2 | 171 | 73 | |
GenBank mRNA | AF011429.1 | - | 1 - 2 | 91 | 73 |
AK292040.1 | - | 1 - 2 | 92 | 73 | |
AY358494.1 | - | 1 - 2 | 125 | 73 | |
BC033586.1 | - | 1 - 2 | 59 | 73 | |
DA957550.1 | - | 1 - 2 | 171 | 73 | |
U82812.1 | - | 1 - 2 | 88 | 73 |