The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005407.1 | NP_001005407.1 | 2 - 3 | 553 | 119 |
NM_021098.2 | NP_066921.2 | 2 - 3 | 553 | 119 | |
XM_005255652.4 | XP_005255709.1 | 2 - 3 | 399 | 119 | |
XM_006720963.3 | XP_006721026.1 | 2 - 3 | 399 | 119 | |
XM_006720964.3 | XP_006721027.1 | 2 - 3 | 399 | 119 | |
XM_006720965.3 | XP_006721028.1 | 2 - 3 | 399 | 119 | |
XM_006720967.3 | XP_006721030.1 | 2 - 3 | 400 | 119 | |
XM_006720968.3 | XP_006721031.1 | 2 - 3 | 400 | 119 | |
XM_011522727.2 | XP_011521029.1 | 2 - 3 | 406 | 119 | |
XM_017023819.1 | XP_016879308.1 | 2 - 3 | 399 | 119 | |
XM_017023820.1 | XP_016879309.1 | 2 - 3 | 399 | 119 | |
XM_017023821.1 | XP_016879310.1 | 2 - 3 | 399 | 119 | |
GenBank mRNA | AF051946.3 | - | 2 - 3 | 384 | 119 |
AF073931.1 | - | 2 - 3 | 553 | 119 | |
AJ420779.1 | - | 1 - 2 | 305 | 119 |