The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024912.2 | NP_001020083.1 | 4 - 5 | 1098 | 78 |
NM_001205344.1 | NP_001192273.1 | 4 - 5 | 1098 | 78 | |
NM_001712.4 | NP_001703.2 | 4 - 5 | 1098 | 78 | |
GenBank mRNA | AK314211.1 | - | 4 - 5 | 1070 | 78 |
AL833584.1 | - | 4 - 5 | 1068 | 78 | |
BC014473.1 | - | 4 - 5 | 1055 | 78 | |
BC024164.1 | - | 4 - 5 | 1051 | 78 | |
D90311.1 | - | 4 - 5 | 1027 | 78 | |
J03858.1 | - | 4 - 5 | 1044 | 78 | |
M72238.1 | - | 4 - 5 | 1058 | 78 | |
S71326.1 | - | 4 - 5 | 963 | 78 | |
X16354.1 | - | 4 - 5 | 1035 | 78 | |
X16356.1 | - | 3 - 4 | 691 | 78 |