The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135599.2 | NP_001129071.1 | 2 - 3 | 1797 | 91 |
GenBank mRNA | AB209842.1 | - | 2 - 3 | 1725 | 91 |
AK295671.1 | - | 2 - 3 | 276 | 91 | |
AK296504.1 | - | 2 - 3 | 430 | 91 | |
M19154.1 | - | 2 - 3 | 896 | 91 |