The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000174.4 | NP_000165.1 | 1 - 2 | 91 | 101 |
XM_005247374.3 | XP_005247431.1 | 3 - 4 | 387 | 101 | |
XM_011512701.1 | XP_011511003.1 | 4 - 5 | 469 | 101 | |
GenBank mRNA | BC030229.1 | - | 1 - 2 | 56 | 101 |
X52997.1 | - | 1 - 2 | 91 | 101 |