The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000564.4 | NP_000555.2 | 12 - 13 | 1818 | 96 |
NM_175726.3 | NP_783853.1 | 11 - 12 | 1749 | 96 | |
GenBank mRNA | AB288089.1 | - | 7 - 8 | 889 | 96 |
AB288090.1 | - | 5 - 6 | 547 | 96 | |
M96652.1 | - | 12 - 13 | 1423 | 96 | |
X61176.1 | - | 10 - 11 | 1235 | 96 |