The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199189.1 | NP_001186118.1 | 2 - 3 | 596 | 127 |
NM_001239.3 | NP_001230.1 | 2 - 3 | 482 | 127 | |
XM_005248627.3 | XP_005248684.1 | 2 - 3 | 320 | 127 | |
XM_005248629.4 | XP_005248686.1 | 2 - 3 | 658 | 127 | |
XM_011543706.2 | XP_011542008.1 | 2 - 3 | 320 | 127 | |
XM_017010014.1 | XP_016865503.1 | 2 - 3 | 622 | 127 | |
GenBank mRNA | AB209342.1 | - | 1 - 2 | 74 | 127 |
AK291720.1 | - | 2 - 3 | 335 | 127 | |
BC005280.1 | - | 2 - 3 | 338 | 127 | |
BC016705.1 | - | 2 - 3 | 333 | 127 | |
BC016823.1 | - | 2 - 3 | 329 | 127 | |
BC022351.1 | - | 2 - 3 | 317 | 127 | |
BP244381.1 | - | 2 - 3 | 473 | 127 | |
BX537673.1 | - | 2 - 3 | 597 | 127 | |
CR407658.1 | - | 2 - 3 | 243 | 127 | |
U11791.1 | - | 2 - 3 | 303 | 127 | |
U12685.1 | - | 2 - 3 | 255 | 127 |