Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KHK
Entrez Gene ID
3795
Gene Name
ketohexokinase
Gene Aliases
-
Chromosome Location
Chr.2: 27086738 - 27100751 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000221.2 NP_000212.1 5 - 6 1082 91
NM_006488.2 NP_006479.1 5 - 6 1082 91
XM_005264294.3 XP_005264351.1 6 - 7 1225 91
XM_005264296.3 XP_005264353.1 5 - 6 1068 91
XM_005264298.3 XP_005264355.1 3 - 4 835 91
XM_017004060.1 XP_016859549.1 6 - 7 1222 91
XM_017004061.1 XP_016859550.1 4 - 5 952 91
GenBank mRNA AK130033.1 - 5 - 6 599 91
AK223075.1 - 5 - 6 599 91
BC006233.2 - 5 - 6 1020 91
BX648873.1 - 3 - 4 349 91
CR456801.1 - 5 - 6 565 91
X78677.1 - 3 - 4 361 91
X78678.1 - 5 - 6 573 91

View Full Product Details