The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256584.1 | NP_001243513.1 | 4 - 5 | 1339 | 76 |
NM_002101.4 | NP_002092.1 | 3 - 4 | 521 | 76 | |
NM_016815.3 | NP_058131.1 | 2 - 3 | 464 | 76 | |
XM_006712460.3 | XP_006712523.1 | 3 - 4 | 389 | 76 | |
GenBank mRNA | AK312032.1 | - | 3 - 4 | 309 | 76 |
BC104246.1 | - | 3 - 4 | 322 | 76 | |
BC104247.1 | - | 3 - 4 | 322 | 76 | |
BC106051.1 | - | 3 - 4 | 353 | 76 | |
CB992018.1 | - | 2 - 3 | 249 | 76 | |
M11802.1 | - | 3 - 4 | 192 | 76 | |
M28335.1 | - | 3 - 4 | 186 | 76 | |
M36284.1 | - | 3 - 4 | 192 | 76 | |
X12496.1 | - | 3 - 4 | 312 | 76 | |
X51973.1 | - | 2 - 3 | 135 | 76 |