Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR3A2
Entrez Gene ID
4995
Gene Name
olfactory receptor family 3 subfamily A member 2
Gene Aliases
OLFRA04, OR17-14, OR17-228, OR228
Chromosome Location
Chr.17: 3276113 - 3386311 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002551.3 NP_002542.3 1 - 1 530 96
XM_017024697.1 XP_016880186.1 6 - 6 1150 96
XM_017024698.1 XP_016880187.1 4 - 4 842 96
GenBank mRNA BC095517.1 - 1 - 1 530 96

Gene Symbol
OR3A1
Entrez Gene ID
4994
Gene Name
olfactory receptor family 3 subfamily A member 1
Gene Aliases
OLFRA03, OR17-40, OR17-82, OR40
Chromosome Location
Chr.17: 3288318 - 3336087 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002550.2 NP_002541.2 1 - 1 96
XM_017024695.1 XP_016880184.1 2 - 2 96
XM_017024696.1 XP_016880185.1 2 - 2 96
GenBank mRNA BC069414.1 - 1 - 1 96
BC096187.3 - 1 - 1 96
BC096188.3 - 1 - 1 96
BC096189.3 - 1 - 1 96
BC096190.1 - 1 - 1 96

Gene Symbol
OR3A3
Entrez Gene ID
8392
Gene Name
olfactory receptor family 3 subfamily A member 3
Gene Aliases
OR17-137, OR17-16, OR17-201, OR3A6, OR3A7, OR3A8P
Chromosome Location
Chr.17: 3420424 - 3421805 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012373.2 NP_036505.2 1 - 1 491 96
XM_011524028.2 XP_011522330.1 2 - 2 501 96
GenBank mRNA BC069415.1 - 1 - 1 491 96
BC108921.2 - 1 - 1 485 96

View Full Product Details