The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135254.1 | NP_001128726.1 | 4 - 5 | 1186 | 73 |
NM_002584.2 | NP_002575.1 | 4 - 5 | 1186 | 73 | |
NM_013945.2 | NP_039236.1 | 4 - 5 | 1180 | 73 | |
GenBank mRNA | BC121165.2 | - | 4 - 5 | 662 | 73 |
BC121166.2 | - | 4 - 5 | 662 | 73 | |
DQ322591.1 | - | 4 - 5 | 671 | 73 | |
X96743.1 | - | 4 - 5 | 1187 | 73 | |
Z35141.1 | - | 4 - 5 | 690 | 73 |