The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003649.2 | NP_003640.2 | 3 - 4 | 370 | 83 |
GenBank mRNA | AB496420.1 | - | 2 - 3 | 282 | 83 |
AK293029.1 | - | 3 - 4 | 376 | 83 | |
AX786391.1 | - | 2 - 3 | 282 | 83 | |
BC032786.1 | - | 3 - 4 | 370 | 83 | |
D89858.1 | - | 2 - 3 | 282 | 83 |