Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ADAM22
- Entrez Gene ID
-
53616
- Gene Name
- ADAM metallopeptidase domain 22
- Gene Aliases
-
ADAM 22, MDC2
- Chromosome Location
-
Chr.7: 87934143 - 88202889 on Build GRCh38
View Full Product Details