The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017490.3 | NP_059672.2 | 1 - 2 | 452 | 67 |
GenBank mRNA | AK290339.1 | - | 1 - 2 | 420 | 67 |
X97630.1 | - | 1 - 2 | 364 | 67 |