The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130831.1 | NP_001124303.1 | 2 - 3 | 281 | 107 |
NM_201432.1 | NP_958836.1 | 2 - 3 | 438 | 107 | |
NM_201433.1 | NP_958839.1 | 2 - 3 | 468 | 107 | |
XM_005256831.3 | XP_005256888.1 | 3 - 4 | 379 | 107 | |
XM_005256832.3 | XP_005256889.1 | 2 - 3 | 249 | 107 | |
XM_005256833.3 | XP_005256890.1 | 2 - 3 | 442 | 107 | |
XM_005256834.3 | XP_005256891.1 | 2 - 3 | 268 | 107 | |
XM_005256835.3 | XP_005256892.1 | 2 - 3 | 212 | 107 | |
XM_011524044.2 | XP_011522346.1 | 2 - 3 | 273 | 107 | |
XM_017025239.1 | XP_016880728.1 | 2 - 3 | 302 | 107 | |
XM_017025240.1 | XP_016880729.1 | 2 - 3 | 171 | 107 | |
XM_017025241.1 | XP_016880730.1 | 2 - 3 | 147 | 107 | |
XM_017025242.1 | XP_016880731.1 | 2 - 3 | 321 | 107 | |
GenBank mRNA | AB007854.1 | - | 2 - 3 | 236 | 107 |
AB209030.1 | - | 1 - 2 | 125 | 107 | |
AK057761.1 | - | 2 - 3 | 393 | 107 | |
AK225165.1 | - | 2 - 3 | 260 | 107 | |
AK292987.1 | - | 2 - 3 | 254 | 107 | |
AK294829.1 | - | 1 - 2 | 229 | 107 | |
AK299574.1 | - | 2 - 3 | 260 | 107 | |
AK307987.1 | - | 3 - 4 | 290 | 107 | |
AK309722.1 | - | 2 - 3 | 289 | 107 | |
AY327406.1 | - | 2 - 3 | 468 | 107 | |
BC001152.1 | - | 2 - 3 | 261 | 107 | |
BC006454.1 | - | 2 - 3 | 438 | 107 | |
BP202727.1 | - | 2 - 3 | 281 | 107 | |
BT006891.1 | - | 1 - 2 | 115 | 107 |