The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113491.1 | NP_001106963.1 | 4 - 5 | 1037 | 69 |
NM_001113492.1 | NP_001106964.1 | 4 - 5 | 1212 | 69 | |
NM_001113493.1 | NP_001106965.1 | 3 - 4 | 1198 | 69 | |
NM_001113494.1 | NP_001106966.1 | 3 - 4 | 1151 | 69 | |
NM_001113495.1 | NP_001106967.1 | 2 - 3 | 851 | 69 | |
NM_001113496.1 | NP_001106968.1 | 2 - 3 | 446 | 69 | |
NM_001293695.1 | NP_001280624.1 | 3 - 4 | 980 | 69 | |
NM_001293696.1 | NP_001280625.1 | 2 - 3 | 262 | 69 | |
NM_001293697.1 | NP_001280626.1 | 2 - 3 | 475 | 69 | |
NM_001293698.1 | NP_001280627.1 | 2 - 3 | 561 | 69 | |
NM_006640.4 | NP_006631.2 | 3 - 4 | 1669 | 69 | |
XM_005256962.1 | XP_005257019.1 | 2 - 3 | 319 | 69 | |
XM_006721643.2 | XP_006721706.1 | 5 - 6 | 1140 | 69 | |
XM_006721644.1 | XP_006721707.1 | 2 - 3 | 362 | 69 | |
XM_011524204.1 | XP_011522506.1 | 5 - 6 | 1237 | 69 | |
XM_011524206.1 | XP_011522508.1 | 3 - 4 | 957 | 69 | |
XM_011524207.1 | XP_011522509.1 | 3 - 4 | 970 | 69 | |
XM_011524208.2 | XP_011522510.1 | 2 - 3 | 228 | 69 | |
XM_017024031.1 | XP_016879520.1 | 3 - 4 | 569 | 69 | |
XM_017024032.1 | XP_016879521.1 | 2 - 3 | 568 | 69 | |
GenBank mRNA | AB023208.1 | - | 3 - 4 | 1151 | 69 |
AF123052.1 | - | 3 - 4 | 1632 | 69 | |
AF142408.1 | - | 3 - 4 | 1114 | 69 | |
AF189712.2 | - | 5 - 6 | 1164 | 69 | |
AF189713.3 | - | 4 - 5 | 1007 | 69 | |
AJ312319.1 | - | 3 - 4 | 1198 | 69 | |
AJ312320.1 | - | 2 - 3 | 240 | 69 | |
AJ312321.1 | - | 3 - 4 | 1669 | 69 | |
AJ312322.1 | - | 3 - 4 | 1140 | 69 | |
AK022493.1 | - | 2 - 3 | 228 | 69 | |
AK056495.1 | - | 2 - 3 | 851 | 69 | |
AK290368.1 | - | 4 - 5 | 1212 | 69 | |
AK299828.1 | - | 2 - 3 | 475 | 69 | |
AK300268.1 | - | 5 - 6 | 871 | 69 | |
AK300270.1 | - | 3 - 4 | 939 | 69 | |
AK303449.1 | - | 2 - 3 | 255 | 69 | |
AK304143.1 | - | 3 - 4 | 1052 | 69 | |
AK316473.1 | - | 2 - 3 | 561 | 69 | |
BC021192.2 | - | 3 - 4 | 892 | 69 | |
BC054004.1 | - | 5 - 6 | 867 | 69 | |
BT007215.1 | - | 3 - 4 | 857 | 69 | |
CN393537.1 | - | 2 - 3 | 254 | 69 | |
DB148645.1 | - | 2 - 3 | 446 | 69 | |
DC410238.1 | - | 2 - 3 | 263 | 69 | |
KU178538.1 | - | 2 - 3 | 212 | 69 | |
KU178539.1 | - | 3 - 4 | 857 | 69 |