The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007052.4 | NP_008983.2 | 2 - 3 | 349 | 108 |
NM_013955.2 | NP_039249.1 | 2 - 3 | 349 | 108 | |
GenBank mRNA | AF127763.2 | - | - | 349 | 108 |
AF166326.1 | - | 2 - 3 | 213 | 108 | |
AF166327.1 | - | 2 - 3 | 213 | 108 | |
AF166328.1 | - | 2 - 3 | 213 | 108 | |
AK292201.1 | - | 2 - 3 | 304 | 108 | |
AK313678.1 | - | 2 - 3 | 258 | 108 | |
BC075014.2 | - | 2 - 3 | 164 | 108 | |
BC075015.2 | - | 2 - 3 | 164 | 108 | |
BX956931.1 | - | 2 - 3 | 319 | 108 |