Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GGT1
Entrez Gene ID
2678
Gene Name
gamma-glutamyltransferase 1
Gene Aliases
CD224, D22S672, D22S732, GGT, GGT 1, GTG
Chromosome Location
Chr.22: 24583750 - 24629005 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013421.2 NP_038265.2 2 - 3 236 144
GenBank mRNA J04131.1 - 2 - 3 178 144

View Full Product Details