The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159996.2 | NP_001153468.1 | 7 - 8 | 1055 | 79 |
NM_001160004.2 | NP_001153476.1 | 11 - 12 | 1775 | 79 | |
NM_001322206.1 | NP_001309135.1 | 8 - 9 | 2433 | 79 | |
NM_013960.4 | NP_039254.1 | 11 - 12 | 1784 | 79 | |
XM_017013371.1 | XP_016868860.1 | 11 - 12 | 1496 | 79 | |
XM_017013372.1 | XP_016868861.1 | 11 - 12 | 1487 | 79 | |
GenBank mRNA | AK293240.1 | - | 7 - 8 | 890 | 79 |
BC144446.1 | - | 11 - 12 | 1323 | 79 | |
EU363510.1 | - | 11 - 12 | 1255 | 79 | |
U02326.1 | - | 11 - 12 | 1375 | 79 |