Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDTC1
Entrez Gene ID
23038
Gene Name
WD and tetratricopeptide repeats 1
Gene Aliases
ADP, DCAF9
Chromosome Location
Chr.1: 27234516 - 27308633 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276252.1 NP_001263181.1 12 - 13 1770 61
NM_015023.4 NP_055838.2 12 - 13 1767 61
XM_011541057.1 XP_011539359.1 12 - 13 1448 61
XM_011541058.2 XP_011539360.1 12 - 13 1715 61
GenBank mRNA AB028960.1 - 7 - 8 909 61
AK001734.1 - 11 - 12 1242 61
AK023101.1 - 3 - 4 309 61
AK023778.1 - 3 - 4 349 61
AK315564.1 - 12 - 13 1654 61
BC032523.1 - 12 - 13 1630 61

View Full Product Details