Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 41263135-41263637 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UCHL1
Entrez Gene ID
7345
Gene Name
ubiquitin C-terminal hydrolase L1
Gene Aliases
HEL-117, HEL-S-53, NDGOA, PARK5, PGP 9.5, PGP9.5, PGP95, Uch-L1
UniGene
Hs.518731
Gene Chromosome Location
Chr.4: 41256881-41268429 on Build GRCh38
Transcripts
NM_004181.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
230547 c.481C>T p.H161Y Chr.4: 41263246-41263246 on Build GRCh38
3735259 c.485T>C p.F162S Chr.4: 41263250-41263250 on Build GRCh38
6325761 c.514C>T p.L172F Chr.4: 41263279-41263279 on Build GRCh38
108758 c.480C>T p.F160F Chr.4: 41263245-41263245 on Build GRCh38
3428503 c.466G>T p.D156Y Chr.4: 41263231-41263231 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1053785092 Chr.4: 41263599-41263599 on Build GRCh38
rs924509607 Chr.4: 41263559-41263559 on Build GRCh38
rs145093131 Chr.4: 41263278-41263278 on Build GRCh38
rs763513865 Chr.4: 41263281-41263281 on Build GRCh38
rs746488910 Chr.4: 41263244-41263244 on Build GRCh38
rs760961273 Chr.4: 41263321-41263321 on Build GRCh38
rs756738860 Chr.4: 41263159-41263159 on Build GRCh38
rs896096169 Chr.4: 41263319-41263319 on Build GRCh38
rs750609906 Chr.4: 41263203-41263203 on Build GRCh38
rs919371936 Chr.4: 41263583-41263583 on Build GRCh38
rs973501236 Chr.4: 41263578-41263578 on Build GRCh38
rs955886508 Chr.4: 41263504-41263504 on Build GRCh38
rs935291745 Chr.4: 41263362-41263362 on Build GRCh38
rs916988680 Chr.4: 41263533-41263533 on Build GRCh38
rs569364205 Chr.4: 41263495-41263495 on Build GRCh38
rs917125455 Chr.4: 41263271-41263271 on Build GRCh38
rs893875824 Chr.4: 41263380-41263380 on Build GRCh38
rs769027517 Chr.4: 41263283-41263283 on Build GRCh38
rs549234368 Chr.4: 41263441-41263441 on Build GRCh38
rs1000356470 Chr.4: 41263415-41263415 on Build GRCh38
rs780645281 Chr.4: 41263169-41263169 on Build GRCh38
rs754972784 Chr.4: 41263388-41263388 on Build GRCh38
rs55901744 Chr.4: 41263511-41263512 on Build GRCh38
rs770405812 Chr.4: 41263251-41263251 on Build GRCh38
rs749419709 Chr.4: 41263216-41263216 on Build GRCh38
rs770192031 Chr.4: 41263267-41263267 on Build GRCh38
rs935926565 Chr.4: 41263570-41263570 on Build GRCh38
rs146239207 Chr.4: 41263519-41263519 on Build GRCh38
rs745473963 Chr.4: 41263266-41263266 on Build GRCh38
rs750700117 Chr.4: 41263304-41263304 on Build GRCh38
rs765165571 Chr.4: 41263322-41263322 on Build GRCh38
rs374340108 Chr.4: 41263284-41263284 on Build GRCh38
rs971232072 Chr.4: 41263546-41263546 on Build GRCh38
rs758378913 Chr.4: 41263234-41263234 on Build GRCh38
rs182750882 Chr.4: 41263531-41263531 on Build GRCh38
rs1053814747 Chr.4: 41263372-41263372 on Build GRCh38
rs779216663 Chr.4: 41263400-41263400 on Build GRCh38
rs752732067 Chr.4: 41263222-41263222 on Build GRCh38
rs1031442823 Chr.4: 41263460-41263460 on Build GRCh38
rs767597442 Chr.4: 41263301-41263301 on Build GRCh38
rs949932642 Chr.4: 41263276-41263276 on Build GRCh38
rs767643318 Chr.4: 41263192-41263192 on Build GRCh38
rs564599222 Chr.4: 41263258-41263258 on Build GRCh38
rs752536111 Chr.4: 41263324-41263324 on Build GRCh38
rs1009480921 Chr.4: 41263435-41263435 on Build GRCh38
rs143313441 Chr.4: 41263393-41263393 on Build GRCh38
rs373569127 Chr.4: 41263411-41263411 on Build GRCh38
rs778225639 Chr.4: 41263193-41263194 on Build GRCh38
rs1015151845 Chr.4: 41263447-41263447 on Build GRCh38
rs190909976 Chr.4: 41263409-41263409 on Build GRCh38
rs954137968 Chr.4: 41263517-41263517 on Build GRCh38
rs748367000 Chr.4: 41263528-41263528 on Build GRCh38
rs758420359 Chr.4: 41263331-41263331 on Build GRCh38
rs937117462 Chr.4: 41263184-41263184 on Build GRCh38
rs377585438 Chr.4: 41263454-41263454 on Build GRCh38
rs1023088354 Chr.4: 41263336-41263336 on Build GRCh38
rs146069195 Chr.4: 41263245-41263245 on Build GRCh38
rs113045732 Chr.4: 41263452-41263452 on Build GRCh38
rs1057384150 Chr.4: 41263303-41263303 on Build GRCh38
rs1057360644 Chr.4: 41263592-41263592 on Build GRCh38
rs1027318538 Chr.4: 41263520-41263520 on Build GRCh38
rs186464888 Chr.4: 41263556-41263556 on Build GRCh38
rs922504193 Chr.4: 41263513-41263514 on Build GRCh38
rs777409628 Chr.4: 41263236-41263236 on Build GRCh38
rs546572233 Chr.4: 41263357-41263357 on Build GRCh38
rs962300888 Chr.4: 41263448-41263448 on Build GRCh38
rs796225428 Chr.4: 41263512-41263513 on Build GRCh38
rs777613388 Chr.4: 41263609-41263609 on Build GRCh38
rs751583368 Chr.4: 41263186-41263186 on Build GRCh38
rs761805743 Chr.4: 41263295-41263295 on Build GRCh38
rs1051023539 Chr.4: 41263396-41263396 on Build GRCh38
rs929443711 Chr.4: 41263589-41263589 on Build GRCh38
rs751472249 Chr.4: 41263334-41263334 on Build GRCh38
rs371799479 Chr.4: 41263212-41263212 on Build GRCh38
rs762150123 Chr.4: 41263191-41263191 on Build GRCh38
rs563254738 Chr.4: 41263490-41263490 on Build GRCh38
rs777587080 Chr.4: 41263333-41263333 on Build GRCh38
rs778805486 Chr.4: 41263218-41263218 on Build GRCh38

View Full Product Details