Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MFN1
Entrez Gene ID
55669
Gene Name
mitofusin 1
Gene Aliases
hfzo1, hfzo2
Chromosome Location
Chr.3: 179347692 - 179394944 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033540.2 NP_284941.2 1 - 2 120 67
GenBank mRNA AB043589.1 - 1 - 2 85 67
AK000700.1 - 1 - 2 77 67
AK314306.1 - 1 - 2 77 67
BC020661.1 - 1 - 2 86 67
BC040557.1 - 1 - 2 120 67

View Full Product Details