The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022893.3 | NP_075044.2 | 4 - 4 | 3918 | 99 |
XM_017004333.1 | XP_016859822.1 | 4 - 4 | 3686 | 99 | |
XM_017004334.1 | XP_016859823.1 | 3 - 3 | 3816 | 99 | |
XM_017004335.1 | XP_016859824.1 | 3 - 3 | 3626 | 99 | |
XM_017004336.1 | XP_016859825.1 | 4 - 4 | 3481 | 99 | |
XM_017004337.1 | XP_016859826.1 | 3 - 3 | 10387 | 99 | |
XM_017004338.1 | XP_016859827.1 | 3 - 3 | 6131 | 99 | |
GenBank mRNA | AJ404611.1 | - | 4 - 4 | 3918 | 99 |
AK001035.1 | - | 1 - 1 | 1791 | 99 |