The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145417.1 | NP_001138889.1 | 2 - 2 | 1539 | 76 |
NM_018133.3 | NP_060603.2 | 2 - 2 | 2340 | 76 | |
XM_005247571.3 | XP_005247628.1 | 2 - 2 | 2074 | 76 | |
XM_006713684.3 | XP_006713747.1 | 2 - 2 | 1557 | 76 | |
XM_011512949.2 | XP_011511251.1 | 2 - 2 | 1644 | 76 | |
GenBank mRNA | AB046805.1 | - | 2 - 2 | 1688 | 76 |
AK001408.1 | - | 1 - 1 | 593 | 76 | |
AK292557.1 | - | 2 - 2 | 76 | ||
AK302491.1 | - | 2 - 2 | 1506 | 76 | |
AL834289.1 | - | 2 - 2 | 1658 | 76 | |
BC032719.1 | - | 1 - 1 | 1129 | 76 | |
BC093764.1 | - | 2 - 2 | 1640 | 76 | |
BC093790.1 | - | 2 - 2 | 1639 | 76 |