The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193421.1 | NP_001180350.1 | 3 - 3 | 5365 | 69 |
NM_173485.5 | NP_775756.3 | 3 - 3 | 5898 | 69 | |
XM_017027641.1 | XP_016883130.1 | 3 - 3 | 5869 | 69 | |
GenBank mRNA | AF230201.1 | - | 1 - 1 | 86 | 69 |
AY926481.1 | - | 1 - 1 | 90 | 69 |