Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TAP2
Entrez Gene ID
6891
Gene Name
transporter 2, ATP binding cassette subfamily B member
Gene Aliases
ABC18, ABCB3, APT2, D6S217E, PSF-2, PSF2, RING11
Chromosome Location
Chr.6: 32821833 - 32838823 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018833.2 NP_061313.2 11 - 12 2056 113
GenBank mRNA AF105151.1 - 11 - 12 2013 113

View Full Product Details