The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018843.3 | NP_061331.2 | 5 - 6 | 641 | 90 |
GenBank mRNA | AF125531.1 | - | 6 - 7 | 614 | 90 |
AK223020.1 | - | 5 - 6 | 595 | 90 | |
AK290848.1 | - | 5 - 6 | 416 | 90 | |
BC027322.1 | - | 4 - 5 | 373 | 90 |