The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282660.1 | NP_001269589.1 | 4 - 5 | 518 | 71 |
NM_001282661.1 | NP_001269590.1 | 7 - 8 | 930 | 71 | |
NM_001282662.1 | NP_001269591.1 | 7 - 8 | 930 | 71 | |
NM_001318139.1 | NP_001305068.1 | 5 - 6 | 655 | 71 | |
NM_020831.4 | NP_065882.1 | 7 - 8 | 930 | 71 | |
XM_005261692.1 | XP_005261749.1 | 6 - 7 | 735 | 71 | |
XM_005261694.1 | XP_005261751.1 | 6 - 7 | 825 | 71 | |
XM_011530283.1 | XP_011528585.1 | 8 - 9 | 1038 | 71 | |
XM_011530285.1 | XP_011528587.1 | 8 - 9 | 949 | 71 | |
XM_017028888.1 | XP_016884377.1 | 7 - 8 | 1013 | 71 | |
XM_017028889.1 | XP_016884378.1 | 3 - 4 | 2013 | 71 | |
GenBank mRNA | AB037859.2 | - | 7 - 8 | 902 | 71 |
AF368061.1 | - | 7 - 8 | 855 | 71 | |
AJ297257.1 | - | 4 - 5 | 512 | 71 | |
AJ297258.1 | - | 7 - 8 | 912 | 71 | |
BC115039.1 | - | 7 - 8 | 896 | 71 | |
BC139730.1 | - | 5 - 6 | 569 | 71 | |
CR456522.1 | - | 4 - 5 | 329 | 71 | |
HG764171.1 | - | 5 - 6 | 655 | 71 |