The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020982.3 | NP_066192.1 | 1 - 1 | 990 | 108 |
GenBank mRNA | AK091002.1 | - | 1 - 1 | 990 | 108 |
BC051870.1 | - | 2 - 2 | 655 | 108 | |
BC065830.1 | - | 1 - 1 | 318 | 108 | |
CD105948.1 | - | 1 - 1 | 317 | 108 |