Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHAT
Entrez Gene ID
1103
Gene Name
choline O-acetyltransferase
Gene Aliases
CHOACTASE, CMS1A, CMS1A2, CMS6
Chromosome Location
Chr.10: 49609095 - 49665104 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020984.3 NP_066264.3 1 - 2 102 113
GenBank mRNA AF305909.1 - 1 - 2 102 113
D82342.1 - 1 - 2 102 113

View Full Product Details