The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020985.3 | NP_066265.3 | 1 - 2 | 92 | 131 |
GenBank mRNA | D82340.1 | - | 1 - 2 | 92 | 131 |