Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PSAT1
Entrez Gene ID
29968
Gene Name
phosphoserine aminotransferase 1
Gene Aliases
EPIP, NLS2, PSA, PSAT, PSATD
Chromosome Location
Chr.9: 78297075 - 78330093 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021154.4 NP_066977.1 7 - 8 1009 119
GenBank mRNA AF113132.1 - 7 - 8 906 119

View Full Product Details