The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021240.3 | NP_067063.1 | 1 - 2 | 490 | 55 |
GenBank mRNA | AF193873.1 | - | 1 - 2 | 71 | 55 |
AJ301581.1 | - | 1 - 2 | 321 | 55 | |
BC113584.1 | - | 1 - 2 | 490 | 55 | |
BC117245.1 | - | 1 - 2 | 490 | 55 |