Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CARD18
Entrez Gene ID
59082
Gene Name
caspase recruitment domain family member 18
Gene Aliases
ICEBERG, UNQ5804, pseudo-ICE
Chromosome Location
Chr.11: 105137721 - 105139734 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021571.3 NP_067546.1 1 - 1 167 96
GenBank mRNA AF208005.1 - 1 - 1 159 96
AI189838.1 - 2 - 2 96
AY358231.1 - 1 - 1 171 96
BC131610.1 - 1 - 1 159 96

View Full Product Details