Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADAM11
Entrez Gene ID
4185
Gene Name
ADAM metallopeptidase domain 11
Gene Aliases
MDC
Chromosome Location
Chr.17: 44759037 - 44781846 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318933.1 NP_001305862.1 26 - 27 2315 94
NM_002390.5 NP_002381.2 26 - 27 2319 94
GenBank mRNA AB009675.1 - 26 - 27 2298 94
AK296512.1 - 26 - 27 2396 94
D17390.1 - 23 - 24 2038 94

View Full Product Details