The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022346.4 | NP_071741.2 | 13 - 14 | 2142 | 109 |
NR_073124.1 | 12 - 13 | 2032 | 109 | ||
XM_011513876.2 | XP_011512178.1 | 12 - 13 | 1941 | 109 | |
XM_017008543.1 | XP_016864032.1 | 13 - 14 | 2032 | 109 | |
XM_017008544.1 | XP_016864033.1 | 12 - 13 | 1897 | 109 | |
GenBank mRNA | AB013299.1 | - | 13 - 14 | 2016 | 109 |
AF235023.1 | - | 13 - 14 | 1945 | 109 | |
AF331796.1 | - | 13 - 14 | 2053 | 109 | |
AK022512.1 | - | 13 - 14 | 1973 | 109 | |
AK023147.1 | - | 12 - 13 | 1723 | 109 | |
AK027511.1 | - | 4 - 5 | 479 | 109 | |
BC000827.1 | - | 2 - 3 | 230 | 109 | |
BC101476.1 | - | 13 - 14 | 1930 | 109 |