Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLF12
Entrez Gene ID
11278
Gene Name
Kruppel like factor 12
Gene Aliases
AP-2rep, AP2REP, HSPC122
Chromosome Location
Chr.13: 73686012 - 74168295 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007249.4 NP_009180.3 4 - 5 890 106
XM_005266251.3 XP_005266308.1 4 - 5 999 106
XM_011534907.2 XP_011533209.1 3 - 4 713 106
XM_011534908.2 XP_011533210.1 3 - 4 713 106
XM_011534909.2 XP_011533211.1 4 - 5 846 106
XM_011534910.2 XP_011533212.1 3 - 4 677 106
XM_011534911.2 XP_011533213.1 3 - 4 755 106
XM_011534912.2 XP_011533214.1 3 - 4 826 106
XM_017020384.1 XP_016875873.1 3 - 4 657 106
XM_017020385.1 XP_016875874.1 4 - 5 930 106
GenBank mRNA AF113122.1 - 4 - 5 864 106
AF161471.1 - 4 - 5 738 106
AF330041.1 - 3 - 4 676 106
AJ243274.1 - 3 - 4 668 106
AK291397.1 - 4 - 5 986 106
BC019680.1 - 3 - 4 695 106
HF546212.1 - 4 - 5 861 106

View Full Product Details