Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TBX3
Entrez Gene ID
6926
Gene Name
T-box 3
Gene Aliases
TBX3-ISO, UMS, XHL
Chromosome Location
Chr.12: 114670254 - 114684164 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016569.3 NP_057653.3 2 - 3 1623 80
GenBank mRNA AF216750.1 - 2 - 3 775 80
BC025258.1 - 2 - 3 1049 80

View Full Product Details